Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012970.2 | NP_001012988.1 | 5 | 470 | ||
NM_001276348.1 | NP_001263277.1 | 5 | 374 | |||
NM_001276349.1 | NP_001263278.1 | 4 | 297 | |||
XM_011544124.2 | XP_011542426.1 | 6 | 1312 | |||
XM_011544125.2 | XP_011542427.1 | 5 | 1062 | |||
XM_011544128.2 | XP_011542430.1 | 5 | 1573 | |||
XM_011544129.2 | XP_011542431.1 | 6 | 1223 | |||
XM_011544130.2 | XP_011542432.1 | 5 | 1144 | |||
XM_017000571.1 | XP_016856060.1 | 6 | 1342 | |||
XM_017000572.1 | XP_016856061.1 | 6 | 1850 | |||
GenBank mRNA | BC054479.1 | - | 5 | 378 | ||
BX099227.1 | - | 4 | 283 | |||
HY055397.1 | - | 5 | 426 |