Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FAM65C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290268.1 | NP_001277197.1 | 22 | 3190 | ||
NM_080829.3 | NP_543019.2 | 22 | 3123 | |||
NR_110890.1 | 22 | 3349 | ||||
XM_005260294.3 | XP_005260351.1 | 23 | 3187 | |||
XM_006723713.3 | XP_006723776.1 | 22 | 3235 | |||
XM_011528578.2 | XP_011526880.1 | 23 | 3215 | |||
XM_011528579.2 | XP_011526881.1 | 23 | 3353 | |||
XM_011528580.2 | XP_011526882.1 | 23 | 3407 | |||
XM_011528581.2 | XP_011526883.1 | 23 | 3391 | |||
XM_011528585.2 | XP_011526887.1 | 12 | 2538 | |||
XM_017027681.1 | XP_016883170.1 | 14 | 2183 | |||
GenBank mRNA | AK299337.1 | - | 22 | 3039 | ||
CA389799.1 | - | 3 | 175 | |||
CD558804.1 | - | 3 | 217 | |||
DC365439.1 | - | 5 | 533 |