Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256054.2 | NP_001242983.1 | 2 | 532 | ||
NM_018325.4 | NP_060795.1 | 2 | 437 | |||
NM_145005.6 | NP_659442.2 | 2 | 454 | |||
GenBank mRNA | AK057806.1 | - | 2 | 432 | ||
AK291425.1 | - | 2 | 427 | |||
AL832229.1 | - | 1 | 394 | |||
AL832467.1 | - | 2 | 393 | |||
BC020851.2 | - | 1 | 374 | |||
BC068445.1 | - | 1 | 386 | 5298741 | ||
BG718065.1 | - | 2 | 443 | |||
BG722125.1 | - | 2 | 430 | |||
BQ068108.1 | - | 543 | ||||
JN681271.1 | - | 2 | 510 |