Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYSND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040273.2 | NP_001035363.1 | 1 | 1029 | ||
NM_173555.3 | NP_775826.2 | 1 | 1029 | |||
GenBank mRNA | BC016840.1 | - | 1 | 483 | ||
BC030242.1 | - | 1 | 945 | 5176098 | ||
BC111501.1 | - | 1 | 968 | 5732356 |