Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP32P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003554.1 | 5 | 4759 | |||
GenBank mRNA | AJ012755.1 | - | 5 | 4313 | ||
AK124785.1 | - | 10 | 4679 |
Gene Symbol: USP32P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY363305.1 | - | 4 | 3218 |