Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRN4CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203422.3 | NP_981967.1 | 2 | 754 | ||
GenBank mRNA | AL834319.1 | - | 2 | 583 | ||
AY358354.1 | - | 2 | 427 | |||
BC053902.1 | - | 2 | 457 | 6165542 |