Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145115.2 | NP_660090.2 | 8 | 1356 | ||
XM_005250194.2 | XP_005250251.1 | 6 | 1128 | |||
XM_011515905.2 | XP_011514207.1 | 8 | 1667 | |||
XM_011515906.2 | XP_011514208.1 | 9 | 1774 | |||
XM_011515907.2 | XP_011514209.1 | 7 | 1611 | |||
XM_011515908.2 | XP_011514210.1 | 8 | 1681 | |||
XM_017011824.1 | XP_016867313.1 | 8 | 1403 | |||
XM_017011825.1 | XP_016867314.1 | 8 | 1708 | |||
XM_017011826.1 | XP_016867315.1 | 9 | 1385 | |||
GenBank mRNA | AK057030.1 | - | 8 | 1333 | ||
AK160369.1 | - | 3 | 1928 | |||
BC069644.1 | - | 5 | 1021 | |||
BC074902.2 | - | 2 | 347 | |||
BC074903.2 | - | 2 | 347 | |||
BC089402.1 | - | 5 | 898 | |||
BC114573.1 | - | 3 | 795 | 40084742 | ||
BC114941.1 | - | 5 | 1015 | 40084741 | ||
BX648158.1 | - | 7 | 2739 |