Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145115.2 | NP_660090.2 | 5 | 739 | ||
XM_005250194.2 | XP_005250251.1 | 3 | 511 | |||
XM_011515905.2 | XP_011514207.1 | 5 | 1050 | |||
XM_011515906.2 | XP_011514208.1 | 6 | 1157 | |||
XM_011515907.2 | XP_011514209.1 | 4 | 994 | |||
XM_011515908.2 | XP_011514210.1 | 5 | 1064 | |||
XM_011515909.2 | XP_011514211.1 | 5 | 1072 | |||
XM_011515910.2 | XP_011514212.1 | 5 | 1073 | |||
XM_017011824.1 | XP_016867313.1 | 5 | 786 | |||
XM_017011825.1 | XP_016867314.1 | 5 | 1091 | |||
XM_017011826.1 | XP_016867315.1 | 6 | 768 | |||
XM_017011827.1 | XP_016867316.1 | 3 | 511 | |||
GenBank mRNA | AK057030.1 | - | 5 | 716 | ||
AK160369.1 | - | 1 | 337 | |||
BC069644.1 | - | 2 | 404 | |||
BC089402.1 | - | 2 | 281 | |||
BC114573.1 | - | 2 | 394 | 40084742 | ||
BC114941.1 | - | 2 | 398 | 40084741 | ||
BX640720.1 | - | 5 | 739 | |||
BX648158.1 | - | 4 | 2122 |