Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101341.1 | NP_001094811.1 | 3, 4 | 897 | ||
GenBank mRNA | AY102071.1 | - | 4, 5 | 445 | ||
BC042093.1 | - | 3, 4 | 897 | |||
KF856952.1 | - | 3, 4 | 991 |