Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243750.1 | NP_001230679.1 | 2 | 243 | ||
NM_181843.2 | NP_862826.1 | 2 | 243 | |||
XM_017017474.1 | XP_016872963.1 | 2 | 422 | |||
GenBank mRNA | AK123561.1 | - | 2 | 227 | ||
BC018644.1 | - | 2 | 236 | 3956297 | ||
CB140633.1 | - | 2 | 243 |