Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM183CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024473.1 | 1 | 1096 | |||
GenBank mRNA | AL833525.1 | - | 1 | 790 | ||
BC028745.1 | - | 1 | 1100 | |||
BC045833.1 | - | 1 | 1095 | |||
BC126344.1 | - | 1 | 1096 |