Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029860.3 | NP_001025031.2 | 2 | 1160 | ||
NR_036491.1 | 2 | 1204 | ||||
XM_011516987.2 | XP_011515289.1 | 2 | 1106 | |||
XM_011516989.2 | XP_011515291.1 | 2 | 1150 | |||
XM_011516990.1 | XP_011515292.1 | 2 | 376 | |||
XM_011516992.2 | XP_011515294.1 | 2 | 1106 | |||
XM_017013358.1 | XP_016868847.1 | 2 | 1106 | |||
GenBank mRNA | AK310518.1 | - | 2 | 730 | ||
BC090892.1 | - | 2 | 1133 |