Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC286297 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046175.1 | 4 | 2151 | |||
GenBank mRNA | AK097152.1 | - | 4 | 2153 |
Gene Symbol: LOC100996643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716911.3 | XP_006716974.1 | 5 | 1692 |