Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006638.2 | NP_001006639.1 | 7 | 512 | ||
NM_001163989.1 | NP_001157461.1 | 7 | 661 | |||
NM_001163990.1 | NP_001157462.1 | 6 | 401 | |||
NM_175738.4 | NP_783865.1 | 7 | 905 | |||
XM_005257288.4 | XP_005257345.1 | 7 | 530 | |||
GenBank mRNA | AK054846.1 | - | 6 | 1341 | ||
AK057069.1 | - | 1 | 56 | |||
AK098068.1 | - | 7 | 512 | |||
AK127073.1 | - | 7 | 1444 | |||
AK290202.1 | - | 7 | 881 | |||
AK296062.1 | - | 7 | 500 | |||
AK296172.1 | - | 6 | 364 | |||
AK298033.1 | - | 5 | 896 | |||
AK303442.1 | - | 7 | 564 | |||
BC016615.1 | - | 7 | 458 | 4520191 | ||
BC040547.1 | - | 7 | 891 | 5287038 |