Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LUZP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009909.3 | NP_001009909.2 | 9 | 956 | ||
NM_001252008.1 | NP_001238937.1 | 9 | 952 | |||
NM_001252010.1 | NP_001238939.1 | 7 | 830 | |||
XM_017017648.1 | XP_016873137.1 | 9 | 869 | |||
GenBank mRNA | AK309077.1 | - | 7 | 605 | ||
AL832641.3 | - | 9 | 868 | |||
BC132829.1 | - | 9 | 748 | 40146851 | ||
BC144177.1 | - | 9 | 744 |