Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT17P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001443.1 | 1 | 297 | |||
GenBank mRNA | AK127974.1 | - | 4 | 1194 | ||
BC040601.1 | - | 3 | 1198 |