Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181426.1 | NP_852091.1 | 3 | 407 | ||
GenBank mRNA | AK304270.1 | - | 3 | 386 | ||
AL122120.1 | - | 9 | 1164 | |||
DC399927.1 | - | 3 | 407 |