Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161834.2 | NP_001155306.2 | 2 | 611 | ||
XM_011515052.1 | XP_011513354.1 | 2 | 836 | |||
XM_011515053.2 | XP_011513355.1 | 2 | 836 | |||
GenBank mRNA | BX092263.1 | - | 2 | 162 |