Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLFN12L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195790.1 | NP_001182719.1 | 1 | 819 | ||
XM_017024005.1 | XP_016879494.1 | 3 | 1785 | |||
XM_017024006.1 | XP_016879495.1 | 8 | 3987 | |||
XM_017024007.1 | XP_016879496.1 | 6 | 3388 | |||
XM_017024008.1 | XP_016879497.1 | 6 | 7570 | |||
XM_017024009.1 | XP_016879498.1 | 5 | 7500 | |||
GenBank mRNA | AK172761.1 | - | 3 | 1620 |