Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD200R1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008784.2 | NP_001008784.2 | 4 | 680 | ||
NM_001199215.1 | NP_001186144.1 | 5 | 749 | |||
XM_011512763.2 | XP_011511065.1 | 6 | 1129 | |||
XM_017006300.1 | XP_016861789.1 | 5 | 506 | |||
GenBank mRNA | AY284976.1 | - | 5 | 749 | ||
AY552790.1 | - | 4 | 680 |