Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006382.3 | NP_006373.2 | 12 | 2217 | ||
GenBank mRNA | BC143291.1 | - | 5 | 493 | ||
BC171723.1 | - | 3 | 255 | |||
BU738380.1 | - | 1 | 560 | |||
U43383.1 | - | 5 | 511 | |||
U65652.1 | - | 5 | 746 |