Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145416.1 | NP_001138888.1 | 2, 3 | 286 | ||
NM_001321112.1 | NP_001308041.1 | 2, 3 | 395 | |||
NM_001321113.1 | NP_001308042.1 | 2, 3 | 205 | |||
NM_001321114.1 | NP_001308043.1 | 2, 3 | 314 | |||
NM_033408.3 | NP_212133.1 | 2, 3 | 405 | |||
NM_198536.2 | NP_940938.1 | 1, 2 | 784 | |||
XM_005259899.2 | XP_005259956.1 | 2, 3 | 491 | |||
GenBank mRNA | AK291979.1 | - | 2, 3 | 265 | ||
AY358949.1 | - | 2, 3 | 226 | |||
BC064948.1 | - | 1, 2 | 525 | 6025162 | ||
BC091472.1 | - | 2, 3 | 213 | 30335220 | ||
BE889078.1 | - | 2, 3 | 405 | |||
BM352837.1 | - | 2, 3 | 164 | |||
BM451378.1 | - | 2, 3 | 291 | |||
BQ639667.1 | - | 2, 3 | 322 | |||
CB994930.1 | - | 2, 3 | 240 |