Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123376.2 | NP_001116848.1 | 1 | 142 | ||
XM_011540071.2 | XP_011538373.1 | 1 | 151 | |||
GenBank mRNA | AB235418.1 | - | 1 | 97 | ||
BC127789.1 | - | 1 | 142 | 40133552 | ||
BC127790.1 | - | 1 | 142 | 40133554 | ||
BX538120.1 | - | 277 |