Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM200C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080209.1 | NP_001073678.1 | 1 | 1823 | ||
XM_005258130.4 | XP_005258187.1 | 3 | 5174 | |||
XM_011525729.2 | XP_011524031.1 | 3 | 7823 |