Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005260731.2 | XP_005260788.1 | 1 | 162 | ||
XM_017027887.1 | XP_016883376.1 | 1 | 127 | |||
XM_017027888.1 | XP_016883377.1 | 1 | 127 |
Gene Symbol: LOC100287166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC004382.1 | - | 1 | 263 |
Gene Symbol: LOC107987276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017028183.1 | XP_016883672.1 | 5 | 1182 |