Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145432.1 | NP_001138904.1 | 1 | 188 | ||
GenBank mRNA | BC006003.1 | - | 1 | 132 | ||
BC032431.1 | - | 1 | 187 | |||
BC040154.1 | - | 1 | 93 | |||
BC046171.2 | - | 1 | 135 | |||
BC062747.1 | - | 1 | 118 | |||
BC086307.1 | - | 1 | 151 | |||
DA733575.1 | - | 1 | 188 |