Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001669.2 | NP_001001669.2 | 10 | 1511 | ||
XM_005268447.4 | XP_005268504.1 | 10 | 1482 | |||
XM_005268448.1 | XP_005268505.1 | 9 | 1385 | |||
XM_006714784.3 | XP_006714847.1 | 11 | 1850 | |||
XM_011537642.2 | XP_011535944.1 | 10 | 1533 | |||
XM_017009460.1 | XP_016864949.1 | 9 | 1624 | |||
XM_017009461.1 | XP_016864950.1 | 8 | 1274 | |||
GenBank mRNA | AB199796.1 | - | 9 | 1458 | ||
AK123597.1 | - | 2 | 151 | |||
AK300355.1 | - | 9 | 1372 | |||
AK300559.1 | - | 10 | 1610 | |||
AK316002.1 | - | 10 | 1581 | |||
BC041325.1 | - | 10 | 1511 | |||
BF508325.1 | - | 2 | 190 |