Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205854.2 | NP_995326.1 | 1, 2 | 151 | ||
GenBank mRNA | AI857998.1 | - | 2 | 294 | ||
AY102070.1 | - | 1, 2 | 126 | |||
AY359057.1 | - | 1, 2 | 129 |