Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUTM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011540122.2 | XP_011538424.1 | 3 | 1241 | ||
XM_017016607.1 | XP_016872096.1 | 3 | 1242 |