Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QRFPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198179.2 | NP_937822.2 | 1 | 111 | ||
XM_017008693.1 | XP_016864182.1 | 1 | 114 | |||
GenBank mRNA | AK309337.1 | - | 1 | 144 | ||
BG169612.1 | - | 1 | 111 | |||
DA461055.1 | - | 1 | 144 |
Set Membership: |
G-protein coupled receptor |