Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301089.1 | NP_001288018.1 | 4 | 548 | ||
NM_014758.2 | NP_055573.2 | 4 | 2562 | |||
XM_005271546.4 | XP_005271603.1 | 4 | 2561 | |||
XM_011542819.2 | XP_011541121.1 | 4 | 2561 | |||
XM_011542820.2 | XP_011541122.1 | 4 | 2561 | |||
XM_011542822.2 | XP_011541124.1 | 4 | 2561 | |||
XM_011542824.2 | XP_011541126.1 | 4 | 1466 | |||
XM_011542825.2 | XP_011541127.1 | 4 | 728 | |||
XM_011542826.2 | XP_011541128.1 | 4 | 545 | |||
XM_017017726.1 | XP_016873215.1 | 4 | 725 | |||
GenBank mRNA | AF395843.1 | - | 4 | 2014 | ||
AK292457.1 | - | 4 | 2584 | |||
AK294731.1 | - | 2 | 207 | |||
AK304809.1 | - | 5 | 568 | |||
AK309202.1 | - | 4 | 694 | |||
AK316414.1 | - | 4 | 512 | |||
AK316505.1 | - | 4 | 505 | |||
BC031620.1 | - | 4 | 2524 | 5172852 | ||
BU431333.1 | - | 3 | 352 | |||
D87443.1 | - | 4 | 2542 | |||
DN990954.1 | - | 1 | 89 |