Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00517 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135286.1 | 2 | 581 | |||
GenBank mRNA | AA029430.1 | - | 1 | 414 | ||
AK125955.1 | - | 2 | 582 |