Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035267.1 | NP_001030344.1 | 3 | 407 | ||
NM_021104.1 | NP_066927.1 | 4 | 292 | |||
GenBank mRNA | AF026844.1 | - | 1 | 253 | ||
BC014383.1 | - | 1 | 254 | |||
BC015984.1 | - | 1 | 253 | |||
BC032611.1 | - | 1 | 244 | |||
BC062462.1 | - | 1 | 235 | |||
BC064907.1 | - | 1 | 235 | |||
BC066328.1 | - | 3 | 319 | |||
BC070252.1 | - | 1 | 250 | |||
BC070253.1 | - | 1 | 249 | |||
BC070254.1 | - | 1 | 250 | |||
BC070255.1 | - | 1 | 249 | |||
BC070483.1 | - | 1 | 216 | |||
BC082759.1 | - | 3 | 347 | |||
BC105279.1 | - | 1 | 253 | |||
BC105286.1 | - | 1 | 254 | |||
BC105296.1 | - | 1 | 254 | |||
BC105794.1 | - | 1 | 253 | |||
BC105795.1 | - | 1 | 253 | |||
BC105799.1 | - | 1 | 254 | |||
BC142666.1 | - | 1 | 233 | |||
BC142726.1 | - | 1 | 233 | |||
Z12962.1 | - | 4 | 292 |
Gene Symbol: LOC107984974 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017025491.1 | XP_016880980.1 | 8 | 1670 |
Gene Symbol: LOC400590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC062632.1 | - | 1 | 429 | 5248762 |