Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00909 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024484.1 | 1 | 471 | |||
GenBank mRNA | AK001853.1 | - | 1 | 217 | ||
BC036588.2 | - | 1 | 460 | |||
BC132930.1 | - | 1 | 245 | |||
BC132956.1 | - | 1 | 245 | |||
DA004415.1 | - | 1 | 153 | |||
DB148466.1 | - | 1 | 471 |