Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRBOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205272.1 | NP_001192201.1 | 1 | 49 | ||
GenBank mRNA | AK130974.1 | - | 1 | 31 | ||
BC062786.1 | - | 1 | 35 | |||
DA207522.1 | - | 1 | 49 | |||
DB458286.2 | - | 1 | 31 |
Gene Symbol: KRBOX1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122033.1 | 1 | 392 | |||
GenBank mRNA | AK057338.1 | - | 1 | 392 |