Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027486.1 | 3 | 244 | |||
GenBank mRNA | AK301746.1 | - | 3 | 245 | ||
BC058890.1 | - | 3 | 216 | |||
EG327356.1 | - | 3 | 365 |