Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00673 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036488.1 | 1 | 448 | |||
NR_137280.1 | 1 | 450 | ||||
NR_137281.1 | 1 | 450 | ||||
GenBank mRNA | AK000477.1 | - | 1 | 448 | ||
KR778895.1 | - | 1 | 448 | |||
KR778896.1 | - | 1 | 448 | |||
KR778900.1 | - | 1 | 448 |
Gene Symbol: LOC284072 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095403.1 | - | 1 | 962 |