Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PSG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130167.1 | NP_001123639.1 | 3 | 786 | ||
NM_001130168.1 | NP_001123640.1 | 2 | 420 | |||
NM_182707.2 | NP_874366.1 | 3 | 786 | |||
XM_011526986.1 | XP_011525288.1 | 3 | 799 | |||
XM_011526987.2 | XP_011525289.1 | 2 | 578 | |||
XM_011526988.2 | XP_011525290.1 | 3 | 769 | |||
GenBank mRNA | AK300230.1 | - | 2 | 419 | ||
AL545004.3 | - | 2 | 403 | |||
BC137500.1 | - | 3 | 786 | 9021500 | ||
BC142628.1 | - | 3 | 774 | 40147510 | ||
CR749812.1 | - | 3 | 785 | |||
M30629.1 | - | 2 | 550 |