Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NBPF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170755.2 | NP_001164226.1 | 5 | 653 | ||
GenBank mRNA | AK097180.1 | - | 5 | 572 |
Gene Symbol: PFN1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003242.3 | 1 | 256 | |||
GenBank mRNA | BC137262.1 | - | 1 | 184 |