Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271466.2 | NP_001258395.1 | 24 | 2956 | ||
XM_011511905.1 | XP_011510207.1 | 24 | 2913 | |||
XM_011511906.1 | XP_011510208.1 | 24 | 2910 | |||
XM_011511907.1 | XP_011510209.1 | 23 | 2814 | |||
XM_011511911.1 | XP_011510213.1 | 25 | 3029 | |||
XM_011511919.2 | XP_011510221.1 | 14 | 2301 | |||
XM_017005018.1 | XP_016860507.1 | 24 | 3171 | |||
XM_017005019.1 | XP_016860508.1 | 24 | 3089 | |||
XM_017005023.1 | XP_016860512.1 | 25 | 3287 | |||
GenBank mRNA | AB058771.1 | - | 14 | 1852 |