Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERFE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291832.1 | NP_001278761.1 | 8 | 1028 | ||
GenBank mRNA | AK094353.1 | - | 4 | 424 | ||
BC047423.1 | - | 4 | 371 | |||
KF984314.1 | - | 8 | 1028 |