Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006721756.2 | XP_006721819.1 | 8 | 3850 |
Gene Symbol: PTCHD1-AS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073010.2 | 12 | 1320 |