Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101417.3 | NP_001094887.1 | 2 | 421 | ||
NM_001101426.3 | NP_001094896.1 | 2 | 421 | |||
XM_006715770.3 | XP_006715833.1 | 2 | 591 | |||
XM_011515499.2 | XP_011513801.1 | 2 | 566 | |||
XM_011515500.2 | XP_011513802.1 | 2 | 566 | |||
XM_011515501.2 | XP_011513803.1 | 2 | 594 | |||
XM_011515508.2 | XP_011513810.1 | 2 | 923 | |||
XM_011515509.2 | XP_011513811.1 | 2 | 1772 | |||
XM_017012575.1 | XP_016868064.1 | 2 | 566 | |||
XM_017012576.1 | XP_016868065.1 | 2 | 578 | |||
GenBank mRNA | DT932748.1 | - | 5 | 502 |