Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: FAM66A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026789.1 | 1 | 426 | |||
GenBank mRNA | AK311040.1 | - | 1 | 427 | ||
DC372067.1 | - | 1 | 425 |
Gene Symbol: FAM66B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027423.1 | 1 | 426 | |||
GenBank mRNA | AK302471.1 | - | 1 | 426 |
Gene Symbol: FAM66E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027424.1 | 1 | 428 |