Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144944.1 | NP_001138416.1 | 4 | 580 | ||
NM_001144945.1 | NP_001138417.1 | 4 | 502 | |||
NM_033546.3 | NP_291024.1 | 4 | 561 | |||
GenBank mRNA | AK291726.1 | - | 4 | 541 | ||
AY320408.1 | - | 4 | 565 | |||
BC004994.1 | - | 4 | 480 | 2906108 | ||
BG696903.1 | - | 4 | 581 | |||
BG703257.1 | - | 4 | 570 | |||
CA307094.1 | - | 1 | 458 | |||
CN302451.1 | - | 4 | 483 | |||
D82058.1 | - | 4 | 494 | |||
U26162.1 | - | 4 | 535 |
Gene Symbol: LOC104968399 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130143.1 | 3 | 655 | |||
NR_130144.1 | 3 | 337 | ||||
NR_130145.1 | 4 | 462 | ||||
GenBank mRNA | AL550084.3 | - | 1 | 274 | ||
BG201476.1 | - | 2 | 102 | |||
BG218162.1 | - | 2 | 111 | |||
BG721448.1 | - | 3 | 339 | |||
BG773124.1 | - | 3 | 653 | |||
HY010228.1 | - | 4 | 453 |
Gene Symbol: MYL12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303047.1 | NP_001289976.1 | 5 | 896 | ||
NM_001303048.1 | NP_001289977.1 | 4 | 539 | |||
NM_001303049.1 | NP_001289978.1 | 4 | 546 | |||
NM_006471.3 | NP_006462.1 | 4 | 813 | |||
GenBank mRNA | AK222569.1 | - | 4 | 489 | ||
AK291145.1 | - | 4 | 509 | |||
BC016372.1 | - | 4 | 488 | 4293343 | ||
BC031972.1 | - | 4 | 813 | 4836883 | ||
BC032748.1 | - | 4 | 484 | 5563221 | ||
BM561281.1 | - | 5 | 613 | |||
BU615599.1 | - | 1 | 433 | |||
CA309747.1 | - | 1 | 432 | |||
CD702288.1 | - | 4 | 569 | |||
D82059.1 | - | 4 | 479 | |||
EU794621.1 | - | 4 | 813 | |||
X54304.1 | - | 4 | 532 |