Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002568.3 | NP_002559.2 | 6, 7 | 1374 | ||
XM_005250861.2 | XP_005250918.1 | 6, 7 | 2285 | |||
GenBank mRNA | AK095194.1 | - | 6, 7 | 1209 | ||
AK130523.1 | - | 6, 7 | 1374 | |||
AK298990.1 | - | 7, 8 | 1090 | |||
AK303120.1 | - | 7, 8 | 1098 | |||
BC015958.2 | - | 7, 8 | 1269 | 3940309 | ||
BC023217.1 | - | 6, 7 | 1353 | |||
BC023520.2 | - | 6, 7 | 1353 | 4123269 | ||
BC041863.1 | - | 6, 7 | 1998 | |||
Y00345.1 | - | 6, 7 | 1363 | |||
Z48501.1 | - | 6, 7 | 795 |