Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016558.3 | NP_057642.1 | 3 | 1009 | ||
NM_033630.2 | NP_361012.2 | 2 | 540 | |||
GenBank mRNA | AF204271.1 | - | 1 | 505 | ||
AF207829.1 | - | 2 | 420 | |||
BC000785.2 | - | 2 | 404 | 3508136 | ||
BC036709.2 | - | 3 | 1377 | 4620185 | ||
BC041022.1 | - | 3 | 1377 | 3830122 | ||
BU190039.1 | - | 2 | 540 | |||
CR457188.1 | - | 1 | 306 |