Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF655 SIRNA Assays ›
Gene Symbol:
ZNF655
NUDCD3
Gene Name:
zinc finger protein 655
NudC domain containing 3
Gene Aliases:
VIK, VIK-1
NudCL
Chromosome Location:
Chr.7: 99557726 - 99576453 on Build GRCh38
Chromosome Location:
Chr.7: 44382366 - 44490875 on Build GRCh38
UniGene ID:
Hs.599798
Hs.488171
Species:
Human
siRNA ID
HSS167153
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF655
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001009958.1
NP_001009958.1
2
264
NM_001009960.1
NP_001009960.1
2
264
NM_001083956.1
NP_001077425.1
2
437
NM_001085366.1
NP_001078835.1
2
437
NM_001085367.1
NP_001078836.1
2
264
NM_001085368.1
NP_001078837.1
2
264
NM_024061.3
NP_076966.1
2
437
NM_138494.2
NP_612503.1
2
437
XM_017012598.1
XP_016868087.1
2
1124
XM_017012599.1
XP_016868088.1
2
282
XM_017012600.1
XP_016868089.1
2
295
XM_017012601.1
XP_016868090.1
2
281
XM_017012602.1
XP_016868091.1
2
1124
XM_017012603.1
XP_016868092.1
2
1357
XM_017012604.1
XP_016868093.1
2
274
XM_017012605.1
XP_016868094.1
2
158
XM_017012606.1
XP_016868095.1
2
295
XM_017012607.1
XP_016868096.1
2
281
XM_017012608.1
XP_016868097.1
2
1123
XM_017012609.1
XP_016868098.1
2
264
XM_017012610.1
XP_016868099.1
2
245
XM_017012611.1
XP_016868100.1
2
1133
XM_017012612.1
XP_016868101.1
2
1130
XM_017012613.1
XP_016868102.1
1
71
GenBank mRNA
AK027114.1
-
2
255
AK057245.1
-
3
163
AK225976.1
-
2
190
AK304785.1
-
2
141
AK314593.1
-
2
280
AK316024.1
-
2
255
AY099353.1
-
2
263
BC000823.1
-
2
207
BC004288.1
-
2
177
BC007378.2
-
2
131
BC037407.1
-
2
227
5752559
BC063478.1
-
2
437
BE293580.1
-
2
264
BI548533.1
-
2
184
BI912391.1
-
2
183
BM829333.1
-
2
291
BX490067.1
-
2
193
DN993232.1
-
2
200
Gene Symbol: NUDCD3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
AK127163.1
-
137
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of G1/S transition of mitotic cell cycle
molecular_function
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qvdz4:80/10.239.165.207:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0