Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF682 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077349.1 | NP_001070817.1 | 4 | 384 | ||
NM_033196.2 | NP_149973.1 | 4 | 427 | |||
XM_017027455.1 | XP_016882944.1 | 3 | 531 | |||
XM_017027456.1 | XP_016882945.1 | 5 | 491 | |||
GenBank mRNA | AK027845.1 | - | 2 | 179 | ||
AK096571.1 | - | 4 | 425 | |||
AK098164.1 | - | 6 | 630 | |||
AK127015.1 | - | 1 | 991 | |||
BC136304.1 | - | 3 | 344 | 9020291 | ||
DA612976.1 | - | 4 | 429 | |||
DA788529.1 | - | 4 | 423 | |||
DB174863.1 | - | 4 | 405 | |||
DR002650.1 | - | 4 | 397 |