Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM217A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011514415.1 | XP_011512717.1 | 1 | 250 | ||
XM_017010482.1 | XP_016865971.1 | 1 | 253 | |||
GenBank mRNA | AK097426.1 | - | 1 | 253 |